Please login to your online account to display your discounted pricing

Alfa Aesar™ Isoquinoline-3-carbonitrile, 99%

$75.95 - $197.39

Chemical Identifiers

Chemical Name or Material Isoquinoline-3-carbonitrile
CAS 26947-41-1
Molecular Formula C10H6N2
Formula Weight 154.17
MDL Number MFCD00075136
Beilstein 114887
Synonym 3-Cyanoisoquinoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1942603

  SDS
alfa aesar™
L1942603
1g Each for $197.39

AAL19426MD

  SDS
alfa aesar™
L19426MD
250mg Each for $75.95
Description & Specifications

Specifications

Formula Weight 154.17
MDL Number MFCD00075136
Beilstein 114887
CAS 26947-41-1
Chemical Name or Material Isoquinoline-3-carbonitrile
Molecular Formula C10H6N2
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Health Hazard 1 Warning
Melting Point 125° to 128°C
Assay 99%
Synonym 3-Cyanoisoquinoline
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Applications
Isoquinoline-3-carbonitrile is employed as analytical standard to study the thermal degradation of 2-chlorobenzylidenemalononitrile (CS riot control agent) at elevated temperatures.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.