Please login to your online account to display your discounted pricing

Alfa Aesar™ Isoquinoline-1-carboxylic acid, 99%

$31.24 - $99.28

Chemical Identifiers

Chemical Name or Material Isoquinoline-1-carboxylic acid
CAS 486-73-7
Molecular Formula C10H7NO2
Formula Weight 173.17
MDL Number MFCD00006901
Beilstein 129177
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0598503

 
alfa aesar™
L0598503
1g Each for $31.24

AAL0598506

 
alfa aesar™
L0598506
5g Each for $99.28
Description & Specifications

Specifications

Formula Weight 173.17
Chemical Name or Material Isoquinoline-1-carboxylic acid
Assay 99%
Molecular Formula C10H7NO2
MDL Number MFCD00006901
CAS 486-73-7
Beilstein 129177
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈165°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.