Please login to your online account to display your discounted pricing

Isoquinolin-3-amine, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB10019CB

 View more versions of this product

Catalog No. BTB10019CB

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 144.18
Quantity 250mg
Assay 97%
Chemical Name or Material Isoquinolin-3-amine
Assay Percent Range 97%
Molecular Formula C9H8N2
CAS 25475-67-6
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material Isoquinolin-3-amine
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H8N2
Formula Weight 144.18