Please login to your online account to display your discounted pricing

Isopropyltriphenylphosphonium Iodide 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  137080250

 View more versions of this product

Catalog No. AC137080250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Isopropyltriphenylphosphonium iodide
Color Slightly yellow to beige-brown
Melting Point 192° to 197°C
Quantity 25g
Name Note 98%
CAS 24470-78-8
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 246-281-5
Physical Form Powder
Infrared Spectrum Authentic
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C21H22IP
Linear Formula (CH3)2CHPPh3+I-
Formula Weight 432.27
MDL Number MFCD00031595
Packaging Glass bottle

Specifications

Chemical Name or Material Isopropyltriphenylphosphonium iodide
Name Note 98%
CAS 24470-78-8
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C21H22IP
Linear Formula (CH3)2CHPPh3+I-
Formula Weight 432.27
MDL Number MFCD00031595
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents