Please login to your online account to display your discounted pricing

Alfa Aesar™ (Isopropylthio)benzene, 98%

$88.05 - $335.57

Chemical Identifiers

Chemical Name or Material (Isopropylthio)benzene
CAS 3019-20-3
Molecular Formula C6H5SCH(CH3)2
Formula Weight 152.26
MDL Number MFCD00039842
Beilstein 1906756
Synonym Isopropyl phenyl sulfide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1712430

 
alfa aesar™
A1712430
250g Each for $335.57

AAA1712418

 
alfa aesar™
A1712418
50g Each for $88.05
Description & Specifications

Specifications

Formula Weight 152.26
Chemical Name or Material (Isopropylthio)benzene
Assay 98%
Molecular Formula C6H5SCH(CH3)2
MDL Number MFCD00039842
CAS 3019-20-3
Beilstein 1906756
Synonym Isopropyl phenyl sulfide
Odor Stench
Boiling Point 105°C (40mmHg)
Quantity 250g