missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isopropylsulfonyl Chloride 95.0+%, TCI America™
Supplier: TCI America I029125G
Specifications
Isopropylsulfonyl Chloride | |
Yellow | |
C3H7ClO2S | |
25 g | |
isopropylsulfonyl chloride, 2-propanesulfonyl chloride, isopropylsulphonyl chloride, isopropyl sulfonyl chloride, ccris 4139, 2-propanesulfonylchloride, isopropanesulfonyl chloride, iso-propylsulfonyl chloride, 2-propane sulfonyl chloride, isopropylsulphonyl choride | |
CC(C)S(=O)(=O)Cl | |
142.597 | |
142.60 | |
Liquid |
10147-37-2 | |
83°C | |
MFCD00007453 | |
3265 | |
DRINJBFRTLBHNF-UHFFFAOYSA-N | |
propane-2-sulfonyl chloride | |
82408 | |
≥95.0% (GC) |
Chemical Identifiers
10147-37-2 | |
142.597 | |
DRINJBFRTLBHNF-UHFFFAOYSA-N | |
82408 | |
CC(C)S(=O)(=O)Cl |
C3H7ClO2S | |
MFCD00007453 | |
isopropylsulfonyl chloride, 2-propanesulfonyl chloride, isopropylsulphonyl chloride, isopropyl sulfonyl chloride, ccris 4139, 2-propanesulfonylchloride, isopropanesulfonyl chloride, iso-propylsulfonyl chloride, 2-propane sulfonyl chloride, isopropylsulphonyl choride | |
propane-2-sulfonyl chloride |
Safety and Handling
TSCA : No