Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopropylphosphonic dichloride, tech. 90%

Chemical Identifiers

Chemical Name or Material Isopropylphosphonic dichloride
CAS 1498-46-0
Purity Grade Technical Grade
Molecular Formula C3H7Cl2OP
Formula Weight 160.97
MDL Number MFCD00603000
UN Number UN3265
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2710903

 
alfa aesar™
H2710903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2710906

 
alfa aesar™
H2710906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 160.97
Chemical Name or Material Isopropylphosphonic dichloride
Purity Grade Technical Grade
Assay 90%
Molecular Formula C3H7Cl2OP
MDL Number MFCD00603000
CAS 1498-46-0
UN Number UN3265
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Quantity 1g

Applications
Isopropylphosphonic dichloride is used as a raw material for the preparation of bis(fluoroalkyl) alkylphosphonates by reacting with fluoroalcohols in the presence of triethylamine as catalyst.

Solubility
Miscible with ethylene glycol and diethyl ether.

Notes
Moisture sensitive. Incompatible with oxidizing agents, water and bases.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.