Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopropyldiphenylphosphine, 99%

$67.35 - $931.18

Chemical Identifiers

Chemical Name or Material Isopropyldiphenylphosphine
CAS 6372-40-3
Molecular Formula C15H17P
Linear Formula (C6H5)2PCH(CH3)2
Formula Weight 228.27
MDL Number MFCD00015028
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2756503

  SDS
alfa aesar™
H2756503
1g Each for $67.35

AAH2756506

  SDS
alfa aesar™
H2756506
5g Each for $229.03

AAH2756514

  SDS
alfa aesar™
H2756514
25g Each for $931.18
Description & Specifications

Specifications

Formula Weight 228.27
Health Hazard 1 Warning
Boiling Point 165°C (11mmHg)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Chemical Name or Material Isopropyldiphenylphosphine
MDL Number MFCD00015028
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 38° to 41°C
CAS 6372-40-3
Flash Point >110°C (230°F)
Assay 99%
Molecular Formula C15H17P
Linear Formula (C6H5)2PCH(CH3)2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.