Please login to your online account to display your discounted pricing

Isopropylboronic acid, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC370880100

 View more versions of this product

Catalog No. AC370880100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 87.91
Physical Form Flakes
Quantity 10g
Color White
Packaging Glass bottle
Recommended Storage Refrigerator +4°C
Linear Formula (CH3)CHB(OH)
Infrared Spectrum Authentic
CAS 80041-89-0
Chemical Name or Material Isopropylboronic acid
MDL Number MFCD01319021
Molecular Formula C3H9BO2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material Isopropylboronic acid
Assay ≥94%
Molecular Formula C3H9BO2
Linear Formula (CH3)CHB(OH)
Formula Weight 87.91
MDL Number MFCD01319021
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

Refrigerator +4°C

SDS & Safety Documents