Please login to your online account to display your discounted pricing

Isopropylboronic acid, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Isopropylboronic acid
Assay ≥94%
Molecular Formula C3H9BO2
Linear Formula (CH3)CHB(OH)
Formula Weight 87.91
MDL Number MFCD01319021
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC370880100

  SDS
thermo fisher scientific
AC370880100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 87.91
Physical Form Flakes
Quantity 10g
Color White
Packaging Glass bottle
Recommended Storage Refrigerator +4°C
Linear Formula (CH3)CHB(OH)
Infrared Spectrum Authentic
CAS 80041-89-0
Chemical Name or Material Isopropylboronic acid
MDL Number MFCD01319021
Molecular Formula C3H9BO2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

Refrigerator +4°C