Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopropylboronic acid, 98%

$38.73 - $426.42

Chemical Identifiers

Chemical Name or Material Isopropylboronic acid
CAS 80041-89-0
Molecular Formula C3H9BO2
Formula Weight 87.91
MDL Number MFCD01319021
Beilstein 1731883
Synonym 2-Propaneboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1996203

  SDS
alfa aesar™
L1996203
1g Each for $38.73

AAL1996206

  SDS
alfa aesar™
L1996206
5g Each for $120.85

AAL1996214

  SDS
alfa aesar™
L1996214
25g Each for $426.42
Description & Specifications

Specifications

Formula Weight 87.91
CAS 80041-89-0
Chemical Name or Material Isopropylboronic acid
Molecular Formula C3H9BO2
MDL Number MFCD01319021
Recommended Storage 2° to 8°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 1731883
Quantity 1g
Synonym 2-Propaneboronic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%

Applications
Isopropylboronic acid as a reagent is involved in copper-promoted cross-coupling, Domino Heck-Suzuki reactions, Suzuki-Miyaura type couple reactions and alkylation-hydride reduction sequence.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C