missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropylamine Hydroiodide, TCI America™

$134.45 - $464.45

Chemical Identifiers

CAS 66735-20-4
Molecular Formula C3H10IN
Molecular Weight (g/mol) 187.024
InChI Key VMLAEGAAHIIWJX-UHFFFAOYSA-N
Synonym Isopropylammonium Iodide
PubChem CID 91972165
IUPAC Name propan-2-amine;hydroiodide
SMILES CC(C)N.I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I09341G View Documents TCI America
I09341G
1g Each for $134.45
 
I09345G View Documents TCI America
I09345G
5g Each for $464.45
 
Specifications

Chemical Identifiers

66735-20-4
187.024
Isopropylammonium Iodide
propan-2-amine;hydroiodide
C3H10IN
VMLAEGAAHIIWJX-UHFFFAOYSA-N
91972165
CC(C)N.I

Specifications

White
66735-20-4
Isopropylammonium Iodide
CC(C)N.I
187.024
187.02
≥97.0% (N,T)
1g
C3H10IN
VMLAEGAAHIIWJX-UHFFFAOYSA-N
propan-2-amine;hydroiodide
91972165
Crystal-Powder at 20°C
Isopropylamine Hydroiodide
Safety and Handling

Safety and Handling

TSCA : No