missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropylamine, 99+%, Alfa Aesar™

Manufacturer:  Alfa Aesar A15044AP*

 View more versions of this product

Catalog No. AAA15044AP



Description

Description

Applications
Isopropylamine acts as a regulating agent for plastics and an additive in the petroleum industry. It is also useful as an intermediate for pharmaceuticals, coating materials, rubber chemicals and other organic products. It also finds application in binary chemical weapons in combination with isopropyl alcohol. Further, it is used as a solvent and to prepare other chemicals.

Solubility
Miscible with water, chloroform, hexanes, acetone and ethanol.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and perchloryl fluoride.
Specifications

Specifications

Isopropylamine
500mL
≥99%
(CH3)2CHNH2
UN1221
14,5209
JJWLVOIRVHMVIS-UHFFFAOYSA-N
propan-2-amine
6363
59.11
0.691
75-31-0
C3H9N
MFCD00008082
605259
isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino
CC(C)N
59.112
CHEBI:15739
≥99%

Chemical Identifiers

75-31-0
59.112
JJWLVOIRVHMVIS-UHFFFAOYSA-N
6363
propan-2-amine
C3H9N
MFCD00008082
isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino
CHEBI:15739
CC(C)N
Safety and Handling

Safety and Handling

GHS H Statement
H224-H315-H319-H335

GHS P Statement
P210-P261-P280-P240-P303+P361+P353-P305+P351+P338-P304+P340-P362-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: ISOPROPYLAMINE

RTECSNumber : NT8400000

TSCA : TSCA

storageNote1 : Air sensitive

Documents