Learn More
Isopropylamine, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 148920100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Isopropylamine | |
0.6900g/mL | |
−37°C | |
98.5% min. (GC) | |
C3H9N | |
(CH3)2CHNH2 | |
04,152 | |
0.69 | |
isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino | |
JJWLVOIRVHMVIS-UHFFFAOYSA-N | |
propan-2-amine | |
6363 | |
CHEBI:15739 | |
99% |
75-31-0 | |
33.0°C to 34.0°C | |
Authentic | |
Plastic drum | |
1.3725 to 1.3755 | |
10L | |
13,164 | |
15,5255 | |
Solubility in water: soluble. Other solubilities: miscible with alcohol and ether | |
CC(C)N | |
59.11 | |
0.3 mPa.s (20°C) | |
59.11 |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Extremely flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
GHS Signal Word: Danger
EINECSNumber : 200-860-9
RUO â Research Use Only