Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopropylamine, 99+%

$23.22 - $28.18

Chemical Identifiers

Chemical Name or Material Isopropylamine
CAS 75-31-0
Molecular Formula C3H9N
Linear Formula (CH3)2CHNH2
Formula Weight 59.11
MDL Number MFCD00008082
UN Number UN1221
Beilstein 605259
Merck Index 14,5209
Synonym 2-Aminopropane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA15044AP

  SDS
alfa aesar
A15044AP*
500mL Each for $28.18

AAA150440F

  SDS
alfa aesar™
A150440F
2500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA15044AE

  SDS
alfa aesar™
A15044AE
100mL Each for $23.22
Description & Specifications

Specifications

Formula Weight 59.11
Quantity 500mL
DOT Information Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: ISOPROPYLAMINE
Assay ≥99%
Merck Index 14,5209
TSCA TSCA
MDL Number MFCD00008082
Health Hazard 2 GHS H Statement
H224-H315-H319-H335
Extremely flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Linear Formula (CH3)2CHNH2
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately
Beilstein 605259
RTECS Number NT8400000
UN Number UN1221
Health Hazard 1 Danger
Density 0.691
Assay Percent Range ≥99%
Storage Note 1 Air sensitive
Chemical Name or Material Isopropylamine
CAS 75-31-0
Molecular Formula (CH3)2CHNH2

Applications
Isopropylamine acts as a regulating agent for plastics and an additive in the petroleum industry. It is also useful as an intermediate for pharmaceuticals, coating materials, rubber chemicals and other organic products. It also finds application in binary chemical weapons in combination with isopropyl alcohol. Further, it is used as a solvent and to prepare other chemicals.

Solubility
Miscible with water, chloroform, hexanes, acetone and ethanol.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and perchloryl fluoride.

Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately

GHS H Statement
H224-H315-H319-H335
Extremely flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Air sensitive