Please login to your online account to display your discounted pricing

Isopropyl trifluoroacetate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Isopropyl trifluoroacetate
Name Note 98%
CAS 400-38-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H7F3O2
Linear Formula CF3CO2CH(CH3)2
Formula Weight 156.11
MDL Number MFCD00013566
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC309090050

  SDS
thermo fisher scientific
309090050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 156.11
Quantity 5g
Color Clear colorless
Density 1.1000g/mL
EINECS Number 206-922-1
Refractive Index 1.3170 to 1.3190 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Chemical Name or Material Isopropyl trifluoroacetate
Molecular Formula C5H7F3O2
CAS 400-38-4
MDL Number MFCD00013566
Assay Percent Range 98%
Name Note 98%
Linear Formula CF3CO2CH(CH3)2
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.