Learn More
Isopropyl trifluoroacetate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1732222
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Isopropyl trifluoroacetate | |
<-78°C | |
73°C | |
C5H7F3O2 | |
MFCD00013566 | |
UN2924 | |
Moisture sensitive | |
ASAXRKSDVDALDT-UHFFFAOYSA-N | |
propan-2-yl 2,2,2-trifluoroacetate | |
78998 | |
97% |
400-38-4 | |
1.108 | |
−8°C (17°F) | |
1.319 | |
100 g | |
1764312 | |
isopropyl trifluoroacetate, isopropyltrifluoroacetate, isopropyl 2,2,2-trifluoroacetate, acetic acid, trifluoro-, 1-methylethyl ester, iso-propyl trifluoroacetate, trifluoroacetic acid isopropyl ester, pubchem12590, acmc-1ctsh, methylethyl 2,2,2-trifluoroacetate | |
CC(C)OC(=O)C(F)(F)F | |
156.10 | |
156.1 |
Chemical Identifiers
400-38-4 | |
156.10 | |
ASAXRKSDVDALDT-UHFFFAOYSA-N | |
78998 | |
CC(C)OC(=O)C(F)(F)F |
C5H7F3O2 | |
MFCD00013566 | |
isopropyl trifluoroacetate, isopropyltrifluoroacetate, isopropyl 2,2,2-trifluoroacetate, acetic acid, trifluoro-, 1-methylethyl ester, iso-propyl trifluoroacetate, trifluoroacetic acid isopropyl ester, pubchem12590, acmc-1ctsh, methylethyl 2,2,2-trifluoroacetate | |
propan-2-yl 2,2,2-trifluoroacetate |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H318-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber : 206-922-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only