Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopropyl propionate, 98%

Chemical Identifiers

Chemical Name or Material Isopropyl propionate
CAS 637-78-5
Molecular Formula CH3CH2CO2CH(CH3)2
Formula Weight 116.16g/mol
MDL Number MFCD00051553
UN Number UN2409
Beilstein 1699921
Synonym Propionic acid isopropyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1905236

 
alfa aesar™
A19052-36
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 108° to 110°C
Quantity 500g
Flash Point 15°C (59°F)
Chemical Name or Material Isopropyl propionate
CAS 637-78-5
Assay 98%
Molecular Formula CH3CH2CO2CH(CH3)2
Formula Weight 116.16g/mol
MDL Number MFCD00051553
UN Number UN2409
Beilstein 1699921
Synonym Propionic acid isopropyl ester
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPYL PROPIONATE
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapour.