missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Palmitate, NF, 90%, Spectrum™ Chemical

IS160, 142-91-6, C19H38O2

Supplier:  Spectrum Chemical Mfg Cor IS1604LTGLCS4

 View more versions of this product

Catalog No. 18603561


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Isopropyl Palmitate, NF is derived from palm oil and is used as a thickening agent and antistatic agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

142-91-6
0.1%
C19H38O2
MFCD00008993
XUGNVMKQXJXZCD-UHFFFAOYSA-N
propan-2-yl hexadecanoate
90%
100%
Amber Glass Bottle
1.435-1.438
4 x 4 L
CCCCCCCCCCCCCCCC(=O)OC(C)C
298.51

Chemical Identifiers

142-91-6
298.51
XUGNVMKQXJXZCD-UHFFFAOYSA-N
CCCCCCCCCCCCCCCC(=O)OC(C)C
C19H38O2
MFCD00008993
propan-2-yl hexadecanoate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions