missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopropyl nitrate, 98%

$103.78 - $422.54

Chemical Identifiers

CAS 1712-64-7
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00007041
InChI Key GAPFWGOSHOCNBM-UHFFFAOYSA-N
Synonym isopropyl nitrate, nitric acid, 1-methylethyl ester, 2-propyl nitrate, nitric acid, isopropyl ester, isopropylnitrate, isopropylester kyseliny dusicne, isopropylester kyseliny dusicne czech, ch3 2chono2, isonite, iso-propylnitrate
PubChem CID 15575
IUPAC Name propan-2-yl nitrate
SMILES CC(C)O[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6024703 View Documents Alfa Aesar™
H6024703
1g Each for $103.78
 
AAH6024706 View Documents Alfa Aesar™
H6024706
5g Each for $422.54
 
Description

Description

Applications
Isopropyl nitrate is used as a diesel cetane improver.

Solubility
Partly miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, reducing agents, flammable substances, organic materials and metal powders.
Specifications

Chemical Identifiers

1712-64-7
105.093
GAPFWGOSHOCNBM-UHFFFAOYSA-N
15575
CC(C)O[N+](=O)[O-]
C3H7NO3
MFCD00007041
isopropyl nitrate, nitric acid, 1-methylethyl ester, 2-propyl nitrate, nitric acid, isopropyl ester, isopropylnitrate, isopropylester kyseliny dusicne, isopropylester kyseliny dusicne czech, ch3 2chono2, isonite, iso-propylnitrate
propan-2-yl nitrate

Specifications

12°C (54°F)
1g
C3H7NO3
MFCD00007041
1701405
GAPFWGOSHOCNBM-UHFFFAOYSA-N
propan-2-yl nitrate
15575
98%
101°C to 102°C
1712-64-7
(CH3)2CHONO2
UN1222
isopropyl nitrate, nitric acid, 1-methylethyl ester, 2-propyl nitrate, nitric acid, isopropyl ester, isopropylnitrate, isopropylester kyseliny dusicne, isopropylester kyseliny dusicne czech, ch3 2chono2, isonite, iso-propylnitrate
CC(C)O[N+](=O)[O-]
105.093
105.09
Isopropyl nitrate
Safety and Handling

Safety and Handling

GHS H Statement
H225-H272-H332
Highly flammable liquid and vapor.
May intensify fire; oxidizer.
Harmful if inhaled.

GHS P Statement
P221-P210-P220-P303+P361+P353-P403+P235-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPYL NITRATE

RTECSNumber : QU8930000

TSCA : TSCA