Learn More
Isopropyl tetradecanoate, 98%, Thermo Scientific Chemicals
$90.05 - $642.62
Chemical Identifiers
CAS | 110-27-0 |
---|---|
Molecular Formula | C17H34O2 |
Molecular Weight (g/mol) | 270.457 |
MDL Number | MFCD00008982 |
InChI Key | AXISYYRBXTVTFY-UHFFFAOYSA-N |
Synonym | isopropyl myristate, isopropyl tetradecanoate, estergel, bisomel, isomyst, promyr, tetradecanoic acid, 1-methylethyl ester, deltyl extra, kesscomir, tegester |
PubChem CID | 8042 |
IUPAC Name | propan-2-yl tetradecanoate |
SMILES | CCCCCCCCCCCCCC(=O)OC(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA18111AE
|
Thermo Scientific Chemicals
A18111AE |
100 mL |
Each for $90.05
|
|
|||||
AAA18111AP
|
Thermo Scientific Chemicals
A18111AP |
500 mL |
Each for $180.58
|
|
|||||
AAA181110F
|
Thermo Scientific Chemicals
A181110F |
2500 mL |
Each for $642.62
|
|
|||||
Description
Isopropyl tetradecanoate is used as a solvent in perfume products and the oil phase in the preparation of formulations. It finds application in eye makeup, nail care products, soaps and detergents, hair care products, shaving products and other skin care products. It behaves as a non-aqueous component of the two-phase mouthwash, Dentyl pH, thereby it removes bacteria from the oral cavity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsopropyl tetradecanoate is used as a solvent in perfume products and the oil phase in the preparation of formulations. It finds application in eye makeup, nail care products, soaps and detergents, hair care products, shaving products and other skin care products. It behaves as a non-aqueous component of the two-phase mouthwash, Dentyl pH, thereby it removes bacteria from the oral cavity.
Solubility
Miscible with alcohol. Immiscible with water and glycerol.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
110-27-0 | |
270.457 | |
AXISYYRBXTVTFY-UHFFFAOYSA-N | |
8042 | |
CCCCCCCCCCCCCC(=O)OC(C)C |
C17H34O2 | |
MFCD00008982 | |
isopropyl myristate, isopropyl tetradecanoate, estergel, bisomel, isomyst, promyr, tetradecanoic acid, 1-methylethyl ester, deltyl extra, kesscomir, tegester | |
propan-2-yl tetradecanoate |
Specifications
110-27-0 | |
191°C to 194°C (20 mm) | |
0.98 | |
MFCD00008982 | |
isopropyl myristate, isopropyl tetradecanoate, estergel, bisomel, isomyst, promyr, tetradecanoic acid, 1-methylethyl ester, deltyl extra, kesscomir, tegester | |
AXISYYRBXTVTFY-UHFFFAOYSA-N | |
propan-2-yl tetradecanoate | |
8042 | |
Liquid |
0.85 | |
152°C (306°F) | |
C17H34O2 | |
100 mL | |
Miscible with alcohol. Immiscible with water and glycerol. | |
CCCCCCCCCCCCCC(=O)OC(C)C | |
270.457 | |
270.46 | |
Isopropyl myristate |
Safety and Handling
EINECSNumber : 203-751-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only