missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropyl Methanesulfonate 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC412840050

 View more versions of this product

Catalog No. AC412840050


 

Specifications Description & Specifications

Specifications

≥98.5%
Liquid
Glass Bottle
1760
104°C
1.1450g/mL
Authentic
926-06-7
99%
99%
138.19
5g
Isopropyl Methanesulfonate
82.0°C (6.0 mmHg)
Clear colorless
1.4180 to 1.4210 (20°C, 589nm)
C4H10O3S
CH3SO3CH(CH3)2
MFCD00047802

Chemical Identifiers

926-06-7
C4H10O3S
138.19
Isopropyl Methanesulfonate, 99%
99%
CH3SO3CH(CH3)2
MFCD00047802
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Suspected of causing cancer.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 213-132-0

RTECSNumber : PB2275000

TSCA : TSCA