missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropyl Methacrylate (stabilized with MEHQ), TCI America™

Manufacturer:  TCI America M0280500ML

 View more versions of this product

Catalog No. M0280500ML



Specifications

Specifications

Isopropyl Methacrylate (stabilized with MEHQ)
Colorless
4655-34-9
MFCD00048290
Methacrylic Acid Isopropyl Ester
CC(C)OC(=O)C(=C)C
128.171
128.17
≥98.0% (GC)
126°C
500mL
C7H12O2
1992
BOQSSGDQNWEFSX-UHFFFAOYSA-N
propan-2-yl 2-methylprop-2-enoate
20769
Clear Liquid at 20°C

Chemical Identifiers

4655-34-9
128.171
BOQSSGDQNWEFSX-UHFFFAOYSA-N
20769
CC(C)OC(=O)C(=C)C
C7H12O2
MFCD00048290
Methacrylic Acid Isopropyl Ester
propan-2-yl 2-methylprop-2-enoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1039

RTECSNumber : OZ5020000

TSCA : Yes

Documents