missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isopropyl Methacrylate (stabilized with MEHQ), TCI America™
$50.42 - $365.14
Chemical Identifiers
CAS | 4655-34-9 |
---|---|
Molecular Formula | C7H12O2 |
Molecular Weight (g/mol) | 128.171 |
MDL Number | MFCD00048290 |
InChI Key | BOQSSGDQNWEFSX-UHFFFAOYSA-N |
Synonym | Methacrylic Acid Isopropyl Ester |
PubChem CID | 20769 |
IUPAC Name | propan-2-yl 2-methylprop-2-enoate |
SMILES | CC(C)OC(=O)C(=C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
M028025ML |
TCI America
M028025ML |
25mL |
Each for $50.42
|
|
M0280500ML |
TCI America
M0280500ML |
500mL |
Each for $365.14
|
|
Chemical Identifiers
4655-34-9 | |
128.171 | |
BOQSSGDQNWEFSX-UHFFFAOYSA-N | |
20769 | |
CC(C)OC(=O)C(=C)C |
C7H12O2 | |
MFCD00048290 | |
Methacrylic Acid Isopropyl Ester | |
propan-2-yl 2-methylprop-2-enoate |
Specifications
126°C | |
25mL | |
C7H12O2 | |
1992 | |
BOQSSGDQNWEFSX-UHFFFAOYSA-N | |
propan-2-yl 2-methylprop-2-enoate | |
20769 | |
Clear Liquid at 20°C | |
Isopropyl Methacrylate (stabilized with MEHQ) |
Colorless | |
4655-34-9 | |
MFCD00048290 | |
Methacrylic Acid Isopropyl Ester | |
CC(C)OC(=O)C(=C)C | |
128.171 | |
128.17 | |
≥98.0% (GC) |
Safety and Handling
EINECSNumber : (2)-1039
RTECSNumber : OZ5020000
TSCA : Yes