Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopropyl isocyanate, 98+%

Chemical Identifiers

Chemical Name or Material Isopropyl isocyanate
CAS 1795-48-8
Molecular Formula (CH3)2CHNCO
Formula Weight 85.11
MDL Number MFCD00002037
UN Number UN2483
Beilstein 969356
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1610503

 
alfa aesar™
L1610503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 85.11
Chemical Name or Material Isopropyl isocyanate
TSCA TSCA
Assay ≥98%
Molecular Formula (CH3)2CHNCO
MDL Number MFCD00002037
CAS 1795-48-8
UN Number UN2483
Beilstein 969356
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H330-H334-H315-H319-H335
Highly flammable liquid and vapor.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Odor Acrid
Boiling Point 74° to 75°C
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ISOPROPYL ISOCYANATE
Flash Point −3°C (26°F)
Quantity 1g

Applications
Isopropyl isocyanate is used as a reagent in the preparation of androgen receptor antagonist for prostate cancer treatment. It is also used in the synthesis of selective dual inhibitor of mammalian target of rapamycin (mTORC1 and mTORC2). Further, it serves as a reagent in the synthesis of O-aryl N-isopropylcarbamates. In addition to this, it is used as a derivatization reagent during the stereoisomeric analysis of secondary alcohols.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Incompatible with water, alcohols, strong bases, amines, acids and strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H330-H334-H315-H319-H335
Highly flammable liquid and vapor.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C