missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Ether, Stabilized, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor I10521LT

 View more versions of this product

Catalog No. 18611142


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

108-20-3
Amber Glass Bottle
0.0001
ACS
1
1 L
0.99
0.0007 meq/g

Chemical Identifiers

108-20-3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.