missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropyl ether, 98+%, pure, stabilized with BHT, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 158960010

 View more versions of this product

Catalog No. AC158960010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Isopropyl ether
0.005% max.
0.7200g/mL
-29°C
98% min. (GC)
C6H14O
[(CH3)2CH]2O
01,362
15,5258
Solubility in water: 9g/L (20°C). Other solubilities: miscible with most common organic solvents
CC(C)OC(C)C
102.18
7914
102.18
Pure
108-20-3,128-37-0
-85.5°C
68°C
Authentic
Glass bottle
1.3670 to 1.369
1 L
0.72
diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
2-propan-2-yloxypropane
5 to 10ppm BHT
0.38 mPa.s (25°C)
98+%
Liquid

Chemical Identifiers

108-20-3
102.18
diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
2-propan-2-yloxypropane
C6H14O
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
7914
CC(C)OC(C)C
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 203-560-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only