Please login to your online account to display your discounted pricing

Isopropyl Ether (Certified), Fisher Chemical


Quantity 500mL
Packaging Glass Bottle

Manufacturer: fisher scientific company  E141500

 View more versions of this product

Catalog No. E141-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Formula Weight 102.18
Boiling Point 68°C
Melting Point −85°C
Quantity 500mL
UN Number 1159
Flash Point −27°C
DOT Information DOT Class 3, : Flammable Liquid
Density .7250g/cm³
Physical Form Liquid
Molecular Formula C6H14O
Vapor Pressure 150mmHg at 25°C
Water 0.2% Max.
Color Clear
Identification Pass test
Specific Gravity Inclusive Between 0.720
ChemAlert Storage Symbol Red
Residue after Evaporation 0.01 % max.
Stabilizer Inclusive Between 90ppm
CAS 128-37-0
Boiling Range 2 °C max., Inclusive of 67 °C
Chemical Name or Material Isopropyl Ether
Cleanup Symbol Solvent
Isopropyl Peroxide 0.05 % max.
MDL Number MFCD00008880
Synonym 2-Isopropoxypropane
Packaging Glass Bottle
Alcohol 0.2 % max.
Linear Formula [(CH3)2CH]2O
CAS 108-20-3

Specifications

Chemical Name or Material Isopropyl Ether
CAS 108-20-3
CAS 128-37-0
Purity Grade Certified Grade
Molecular Formula C6H14O
Linear Formula [(CH3)2CH]2O
Formula Weight 102.18
MDL Number MFCD00008880
Synonym 2-Isopropoxypropane
Accessories
   

SDS & Safety Documents