Please login to your online account to display your discounted pricing

Isopropyl-β-D-thiogalactopyranoside, ≥99%, for biochemistry, contains ca. 15% 1,4-Dioxane, Acros Organics


Quantity 500mg
Packaging Glass bottle

Manufacturer: fisher scientific acros  228565000

 View more versions of this product

Catalog No. AC228565000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Crystalline Powder or Needles
Formula Weight 238.3
Quantity 500mg
Color White to off-white
EINECS Number 206-703-0
TSCA TSCA
Purity Grade For biochemistry
Specific Rotation Condition − 27.00 (20.00°C c=1,H2O)
Beilstein 17, IV, 3726
Merck Index 15, 5126
Specific Rotation -25° to -33° (20°C, 589nm) (c=1, water)
Appearance of Solution (1% aq. soln.) clear, colorless
Water 0.5% max. (K.F.)
CAS 123-91-1
Chemical Name or Material Isopropyl-β-D-thiogalactopyranoside
Name Note 99+%
Infrared Spectrum Authentic
Assay Percent Range 99+%
Loss on Drying 15% to 16% (90°C, 3 hrs) (vacuum)
Molecular Formula C9H18O5S
CAS 367-93-1
Synonym IPTG
Packaging Glass bottle
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Gently wash with plenty of soap and water.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Isopropyl-β-D-thiogalactopyranoside
Name Note 99+%
CAS 367-93-1
CAS 123-91-1
Purity Grade For biochemistry
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C9H18O5S
Formula Weight 238.3
Synonym IPTG
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Gently wash with plenty of soap and water.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.

SDS & Safety Documents