missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropyl-β-D-thiogalactopyranoside, ≥99%, for biochemistry, contains ca. 15% 1,4-Dioxane, Acros Organics

Chemical Identifiers

CAS 367-93-1,123-91-1
Assay Percent Range 99+%
Molecular Formula C9H18O5S
Formula Weight 238.3
Synonym IPTG
Chemical Name or Material Isopropyl-β-D-thiogalactopyranoside, 99+%
Assay ≥99.0%
Grade For biochemistry
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC228565000 SDS Fisher Scientific Acros
228565000
Glass bottle 500mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC228560025 SDS Fisher Scientific Acros
228560025
Glass bottle 2.5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

367-93-1,123-91-1
C9H18O5S
IPTG
≥99.0%
99+%
238.3
Isopropyl-β-D-thiogalactopyranoside, 99+%
For biochemistry

Specifications

− 27.00 (20.00°C c=1,H2O)
500mg
(1% aq. soln.) clear, colorless
Authentic
15% to 16% (90°C, 3 hrs) (vacuum)
238.3
15, 5126
Glass bottle
0.5% max. (K.F.)
≥99.0%
White to off-white
367-93-1,123-91-1
Crystalline Powder or Needles
99+%
C9H18O5S
17, IV, 3726
IPTG
-25° to -33° (20°C, 589nm) (c=1, water)
Isopropyl-β-D-thiogalactopyranoside, 99+%
For biochemistry

Safety and Handling

GHS H Statement:
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Gently wash with plenty of soap and water.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 206-703-0

TSCA : TSCA