Please login to your online account to display your discounted pricing

Isopropyl cinnamate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Isopropyl cinnamate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H14O2
Linear Formula C6H5CHCHCOOCH(CH3)2
Formula Weight 190.24
MDL Number MFCD00048292
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC381870100

  SDS
thermo fisher scientific
AC381870100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 190.24
Quantity 10g
Color Clear yellow
Chemical Name or Material Isopropyl cinnamate
Molecular Formula C12H14O2
Packaging Glass bottle
Assay Percent Range 98%
MDL Number MFCD00048292
CAS 7780-06-5
Refractive Index 1.5450 to 1.5470 (20°C, 589nm)
Infrared Spectrum Authentic
Linear Formula C6H5CHCHCOOCH(CH3)2