Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopropyl cinnamate, 98%

$38.18 - $91.73

Chemical Identifiers

Chemical Name or Material Isopropyl cinnamate
CAS 7780-06-5
Molecular Formula C12H14O2
Formula Weight 190.24
MDL Number MFCD00048292
Beilstein 1908938
Synonym Cinnamic acid isopropyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0782414

 
alfa aesar™
L0782414
25g Each for $38.18

AAL0782422

 
alfa aesar™
L0782422
100g Each for $91.73
Description & Specifications

Specifications

Formula Weight 190.24
Chemical Name or Material Isopropyl cinnamate
Assay 98%
Molecular Formula C12H14O2
MDL Number MFCD00048292
CAS 7780-06-5
Synonym Cinnamic acid isopropyl ester
Flash Point 94°C (201°F)
TSCA TSCA
Beilstein 1908938
RTECS Number GD9625000
Boiling Point 268° to 270°C
Quantity 25g