Please login to your online account to display your discounted pricing

Isopropyl Bromoacetate 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Isopropyl Bromoacetate
Name Note 98%
CAS 29921-57-1
Assay ≥97.5%
Assay Percent Range 97%
Molecular Formula C5H9BrO2
Linear Formula BrCH2CO2CH(CH3)2
Formula Weight 181.03
MDL Number MFCD00009886
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC412811000

  SDS
thermo fisher scientific
AC412811000
Glass Bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥97.5%
Formula Weight 181.03
Quantity 100g
Chemical Name or Material Isopropyl Bromoacetate
Packaging Glass Bottle
Density 1.3990g/mL
Flash Point >110°C
Boiling Point 59.0° to 61.0°C (10.0mmHg)
Color Clear colorless to yellow
EINECS Number 249-956-2
TSCA TSCA
Health Hazard 1 Warning
Refractive Index 1.4440 to 1.4460 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Name Note 98%
Infrared Spectrum Authentic
Assay Percent Range 97%
MDL Number MFCD00009886
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C5H9BrO2
Linear Formula BrCH2CO2CH(CH3)2
CAS 29921-57-1
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.