Please login to your online account to display your discounted pricing

Isopropyl-beta-D-thiogalactopyranoside, 99+%, for mol biology, DNAse, RNAse an, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  327280050

 View more versions of this product

Catalog No. AC327280050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Crystalline Powder
Formula Weight 238.31
Quantity 5g
Color White
Purity Grade For molecular biology
Specific Rotation Condition − 31.00 (20.00°C c=1,water)
Chemical Name or Material Isopropyl-β-D-thiogalactopyranoside
DNase RNase Protease act none detected
Assay Percent Range 99+%
Molecular Formula C9H18O5S
MDL Number MFCD00063273
CAS 367-93-1
Water 1% max. (K.F.)
Infrared Spectrum Authentic
Synonym IPTG
Packaging Glass bottle
Health Hazard 1
Dioxane (GC) none detected
Name Note DNAse, RNAse and Protease Free
Specific Rotation -29° to -33° (20°C, 589nm) (c=1, H2O)

Specifications

Chemical Name or Material Isopropyl-β-D-thiogalactopyranoside
Name Note DNAse, RNAse and Protease Free
CAS 367-93-1
Purity Grade For molecular biology
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C9H18O5S
Formula Weight 238.31
MDL Number MFCD00063273
Synonym IPTG
Accessories

SDS & Safety Documents