missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropyl-beta-D-thiogalactopyranoside, 99+%, for mol biology, DNAse, RNAse an, ACROS Organics™

Chemical Identifiers

CAS 367-93-1
Assay Percent Range 99+%
Molecular Formula C9H18O5S
Formula Weight 238.31
MDL Number MFCD00063273
Synonym IPTG
Chemical Name or Material Isopropyl-β-D-thiogalactopyranoside, DNAse, RNAse and Protease Free
Assay ≥99.0%
Grade For molecular biology
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC327280010 SDS Fisher Scientific Acros
AC327280010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC327280050 SDS Fisher Scientific Acros
327280050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC327280250 SDS Thermo Fisher Scientific
AC327280250
Glass Bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

367-93-1
C9H18O5S
MFCD00063273
Isopropyl-β-D-thiogalactopyranoside, DNAse, RNAse and Protease Free
For molecular biology
99+%
238.31
IPTG
≥99.0%

Specifications

− 31.00 (20.00°C c=1,water)
1g
(GC) none detected
Crystalline Powder
99+%
238.31
Glass bottle
-29° to -33° (20°C, 589nm) (c=1, H2O)
Isopropyl-β-D-thiogalactopyranoside, DNAse, RNAse and Protease Free
For molecular biology
White
367-93-1
none detected
Authentic
C9H18O5S
MFCD00063273
IPTG
1% max. (K.F.)
≥99.0%

Safety and Handling