Please login to your online account to display your discounted pricing

Isopropyl-beta-D-thiogalactopyranoside, 99+%, for mol biology, DNAse, RNAse an, ACROS Organics™


Quantity 25g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC327280250

 View more versions of this product

Catalog No. AC327280250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥99.0%
Formula Weight 238.31
Quantity 25g
Packaging Glass Bottle
Melting Point 105.0°C
Color White
Purity Grade For molecular biology
Specific Rotation Condition − 31.00 (20.00°C c=1,water)
Synonym IPTG
Specific Rotation -29° to -33° (20°C, 589nm) (c=1, H2O)
Water 1% max. (K.F.)
Chemical Name or Material Isopropyl-β-D-thiogalactopyranoside
DNase RNase Protease act none detected
Name Note DNAse, RNAse and Protease Free
Infrared Spectrum Authentic
Assay Percent Range 99+%
MDL Number MFCD00063273
CAS 367-93-1
Health Hazard 1
Dioxane (GC) none detected
Molecular Formula C9H18O5S

Specifications

Chemical Name or Material Isopropyl-β-D-thiogalactopyranoside
Name Note DNAse, RNAse and Protease Free
CAS 367-93-1
Purity Grade For molecular biology
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C9H18O5S
Formula Weight 238.31
MDL Number MFCD00063273
Synonym IPTG
Accessories

SDS & Safety Documents