missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropyl-beta-D-thiogalactopyranoside, 99%, for biochemistry, dioxane-free, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 302790100

 View more versions of this product

Catalog No. AC302790100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Isopropyl-β-D-thiogalactopyranoside
White
99%
(5% aq. soln.) clear, colorless
Powder or Flakes
≥98.5%
C9 H18 O5 S
MFCD00063273
15, 5126
Glass bottle
1% max. (K.F.)
− 31.00 (20.00°C c=1,water)
10g
367-93-1
For biochemistry
Authentic
99%
238.31
17, IV, 3726
IPTG
-29° to -33° (20°C, 589nm) (c=1, water)

Chemical Identifiers

367-93-1
C9 H18 O5 S
MFCD00063273
Isopropyl-β-D-thiogalactopyranoside, 99%
For biochemistry
99%
238.31
IPTG
≥98.5%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 206-703-0

TSCA : TSCA