Please login to your online account to display your discounted pricing

Isopropyl-beta-D-thiogalactopyranoside, 99%, for biochemistry, dioxane-free, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Isopropyl-β-D-thiogalactopyranoside
Name Note 99%
CAS 367-93-1
Purity Grade For biochemistry
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9 H18 O5 S
Formula Weight 238.31
MDL Number MFCD00063273
Synonym IPTG
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC302790010

  SDS
fisher scientific acros
302790010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC302790100

  SDS
fisher scientific acros
302790100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC302790250

  SDS
fisher scientific acros
AC302790250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Powder or Flakes
Formula Weight 238.31
Quantity 1g
Color White
Beilstein 17, IV, 3726
Merck Index 15, 5126
TSCA TSCA
EINECS Number 206-703-0
Purity Grade For biochemistry
Specific Rotation Condition − 31.00 (20.00°C c=1,water)
Appearance of Solution (5% aq. soln.) clear, colorless
Synonym IPTG
Infrared Spectrum Authentic
Packaging Glass bottle
Water 1% max. (K.F.)
Specific Rotation -29° to -33° (20°C, 589nm) (c=1, water)
Health Hazard 1
Molecular Formula C9 H18 O5 S
Name Note 99%
Assay Percent Range 99%
Chemical Name or Material Isopropyl-β-D-thiogalactopyranoside
CAS 367-93-1
MDL Number MFCD00063273
Safety and Handling