missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropyl Alcohol USP, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 67-63-0
Molecular Formula CH3CHOHCH3
Formula Weight 60.1
Chemical Name or Material Isopropyl Alcohol
Grade BP/EP/FCC/JP/NF/USP
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK30314 Avantor Performance
303108
Glass 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

67-63-0
60.1
BP/EP/FCC/JP/NF/USP
CH3CHOHCH3
Isopropyl Alcohol

Specifications

67-63-0
60.1
4L
BP/EP/FCC/JP/NF/USP
CH3CHOHCH3
Glass
Isopropyl Alcohol