Please login to your online account to display your discounted pricing

Isopropyl Alcohol, U.S.P., J.T.Baker™

$73.40 - $1,522.08

Chemical Identifiers

Chemical Name or Material Isopropyl Alcohol
CAS 67-63-0
Purity Grade USP
Molecular Formula CH3CHOHCH3
Formula Weight 60.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003132

 
avantor performance
908003
Narrow Mouth Amber Glass Bottle 4L Each for $1,522.08

02003133

 
avantor performance
908001
Narrow Mouth Amber Glass Bottle 500mL Each for $73.40
Description & Specifications

Specifications

Formula Weight 60.1
CAS 67-63-0
Chemical Name or Material Isopropyl Alcohol
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.79kg
Purity Grade USP
Flash Point 12.00°C
Quantity 4L
Molecular Formula CH3CHOHCH3