Please login to your online account to display your discounted pricing

Isopropyl Alcohol, Multi-Compendial, U.S.P., J.T.Baker™

$195.27 - $1,405.97

Chemical Identifiers

Chemical Name or Material Isopropyl Alcohol
CAS 67-63-0
Purity Grade Multi-Compendial/USP
Molecular Formula CH3CHOHCH3
Formula Weight 60.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003136

 
avantor performance
903703
Narrow Mouth Amber Glass Bottle 4L Each for $1,405.97

02003137

 
avantor performance
903701
Narrow Mouth Amber Glass Bottle 500mL Each for $195.27
Description & Specifications

Specifications

Purity Grade Multi-Compendial/USP
Formula Weight 60.1
Molecular Formula CH3CHOHCH3
CAS 67-63-0
Chemical Name or Material Isopropyl Alcohol
Flash Point 12.00°C
Quantity 4L
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.79kg