missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropyl Alcohol 99.5+%, TCI America™

$31.27 - $55.95

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym 2-hydroxypropane, 2-propanol, alkolave, avantine, dimethylcarbinol, hartosol, isopropanol, isopropyl alcohol, petrohol, sec-propyl alcohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I0164100ML View Documents Tci America
I0164100ML
100mL Each for $31.27
 
I0164500ML View Documents Tci America
I0164500ML
500mL Each for $55.95
 
Specifications

Chemical Identifiers

67-63-0
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
C3H8O
MFCD00011674
2-hydroxypropane, 2-propanol, alkolave, avantine, dimethylcarbinol, hartosol, isopropanol, isopropyl alcohol, petrohol, sec-propyl alcohol
CHEBI:17824
CC(C)O

Specifications

82°C
100mL
C3H8O
1219
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.10
≥99.5% (GC)
Colorless
67-63-0
MFCD00011674
2-hydroxypropane, 2-propanol, alkolave, avantine, dimethylcarbinol, hartosol, isopropanol, isopropyl alcohol, petrohol, sec-propyl alcohol
CC(C)O
60.096
CHEBI:17824
Clear Liquid at 20°C
Isopropyl Alcohol [for Spectrophotometry]
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0207

RTECSNumber : NT8050000

TSCA : Yes