Please login to your online account to display your discounted pricing

Isopropyl Alcohol ChromAR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Isopropyl Alcohol
CAS 67-63-0
Molecular Formula CH3CHOHCH3
Formula Weight 60.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK30431

 
avantor performance
304306
Glass 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK304320

 
avantor performance
304323
NOWPak 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK30434

 
avantor performance
304310
Glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Isopropyl Alcohol
CAS 67-63-0
Molecular Formula CH3CHOHCH3
Formula Weight 60.1
Packaging Glass
Quantity 1L