Please login to your online account to display your discounted pricing

Isopropyl Alcohol, AR, For ACS analysis, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Isopropyl Alcohol
Name Note ACS
CAS 67-63-0
Purity Grade ACS Reagent Grade
Physical Form Liquid
Assay Percent Range 99.5 % min.
Molecular Formula CH3CHOHCH3
Formula Weight 60.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

14650371

 
avantor performance
303221
20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK30321

 
avantor performance
303206
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 20L
Purity Grade ACS Reagent Grade
Physical Form Liquid
Formula Weight 60.1
Color Clear colorless
Chemical Name or Material Isopropyl Alcohol
Molecular Formula CH3CHOHCH3
Assay Percent Range 99.5 % min.
Name Note ACS
CAS 67-63-0