Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopropyl acetoacetate, 98%

$50.34 - $50.34

Chemical Identifiers

Chemical Name or Material Isopropyl acetoacetate
CAS 542-08-5
Molecular Formula C7H12O3
Formula Weight 144.17
MDL Number MFCD00043933
Beilstein 1757046
Merck Index 14,5206
Synonym Acetoacetic acid isopropyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1190718

 
alfa aesar™
A1190718
50g Each for $50.34
Description & Specifications

Specifications

Formula Weight 144.17
CAS 542-08-5
Chemical Name or Material Isopropyl acetoacetate
TSCA TSCA
Assay 98%
Molecular Formula C7H12O3
MDL Number MFCD00043933
Beilstein 1757046
Merck Index 14,5206
Synonym Acetoacetic acid isopropyl ester
Solubility Information Slightly soluble in water
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 73°C (163°F)
Boiling Point 185° to 187°C
Melting Point −27°C
Quantity 50g

Applications
Isopropyl acetoacetate is used as an intermediate for the manufacturing of agrochemicals and pharmaceutical products such as isradipine.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.