Learn More
Isopropyl acetate, 99+%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 180800025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Isopropyl acetate | |
-73.0°C | |
0.8720g/mL | |
4°C | |
99% min. (GC) | |
C5H10O2 | |
CH3CO2CH(CH3)2 | |
2.5 L | |
0.01% max. | |
15, 5251 | |
Solubility in water: 31g/L (20°C). Other solubilities: soluble in most common organic solvents | |
CC(C)OC(=O)C | |
102.13 | |
0.49 mPa.s (25°C) | |
99+% | |
Liquid |
108-21-4 | |
Colorless | |
88.8°C | |
Authentic | |
Glass bottle | |
1.3760 to 1.3780 | |
MFCD00008877 | |
02, 130 | |
0.872 | |
isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester | |
JMMWKPVZQRWMSS-UHFFFAOYSA-N | |
propan-2-yl acetate | |
7915 | |
102.13 | |
Pure |
Chemical Identifiers
108-21-4 | |
JMMWKPVZQRWMSS-UHFFFAOYSA-N | |
7915 | |
CC(C)OC(=O)C |
MFCD00008877 | |
isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester | |
propan-2-yl acetate |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 203-561-1
RUO – Research Use Only