missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropyl acetate, +99%, pure, ACROS Organics™

Manufacturer:  Acros Organics 180801000

 View more versions of this product

Catalog No. AC180801000



Specifications

Specifications

Isopropyl acetate
4°C
1.3760 to 1.3780
0.872
88.8°C
-73.0°C
108-21-4
99+%
CH3CO2CH(CH3)2
02, 130
isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester
JMMWKPVZQRWMSS-UHFFFAOYSA-N
propan-2-yl acetate
7915
Liquid
0.8720g/mL
Authentic
0.01% max.
0.1% max.
Colorless
100mL
Pure
C5H10O2
MFCD00008877
15, 5251
Solubility in water: 31g/L (20°C). Other solubilities: soluble in most common organic solvents
CC(C)OC(=O)C
102.133
102.13
≥99%

Chemical Identifiers

108-21-4
102.133
JMMWKPVZQRWMSS-UHFFFAOYSA-N
7915
CC(C)OC(=O)C
C5H10O2
MFCD00008877
isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester
propan-2-yl acetate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents