missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropyl acetate, 99%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 150860250

 View more versions of this product

Catalog No. AC150860250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Isopropyl acetate
108-21-4
Colorless
88.8°C
Authentic
Metal Drum
1.3760 to 1.3780
MFCD00008877
02, 130
15, 5251
Solubility in water: 31g/L (20°C). Other solubilities: soluble in most common organic solvents
CC(C)OC(=O)C
102.13
0.49 mPa.s (25°C)
99%
Liquid
99%
-73.0°C
0.8720g/mL
4°C
98.5% min. (GC)
C5H10O2
CH3CO2CH(CH3)2
25 L
0.872
isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester
JMMWKPVZQRWMSS-UHFFFAOYSA-N
propan-2-yl acetate
7915
102.13
Pure

Chemical Identifiers

108-21-4
102.13
JMMWKPVZQRWMSS-UHFFFAOYSA-N
7915
CC(C)OC(=O)C
C5H10O2
MFCD00008877
isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester
propan-2-yl acetate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-561-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only