Please login to your online account to display your discounted pricing

Alfa Aesar™ Isopropyl 4,4,4-trifluoroacetoacetate, 95%

$124.68 - $124.68

Chemical Identifiers

Chemical Name or Material Isopropyl 4,4,4-trifluoroacetoacetate
CAS 175230-50-9
Molecular Formula C7H9F3O3
Formula Weight 198.14
MDL Number MFCD00040990
Beilstein 7475563
Synonym 4,4,4-Trifluoroacetoacetic acid isopropyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1019314

 
alfa aesar™
L1019314
25g Each for $124.68
Description & Specifications

Specifications

Formula Weight 198.14
CAS 175230-50-9
Chemical Name or Material Isopropyl 4,4,4-trifluoroacetoacetate
Assay 95%
MDL Number MFCD00040990
Boiling Point 42°C (28mmHg)
Molecular Formula C7H9F3O3
Beilstein 7475563
Synonym 4,4,4-Trifluoroacetoacetic acid isopropyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.