missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropyl 4,4,4-trifluoroacetoacetate, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals L1019314

 View more versions of this product

Catalog No. AAL1019314



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Isopropyl 4,4,4-trifluoroacetoacetate
42°C (28 mmHg)
1.3891
25 g
isopropyl 4,4,4-trifluoroacetoacetate, isopropyl 4,4,4-trifluoro-3-oxobutanoate, isopropyl 4,4,4-trifluoro-3-oxobutyrate, isopropyl-4,4,4-trifluoroacetoacetate, acmc-209e9i, isopropyl4,4,4-trifluoroacetoacetate, methylethyl 4,4,4-trifluoro-3-oxobutanoate, isopropyl 4,4,4-trifluoro-3-oxobutanoate #, 4,4,4-trifluoroacetoacetic acid isopropyl ester
CC(C)OC(=O)CC(=O)C(F)(F)F
198.14
198.14
175230-50-9
C7H9F3O3
MFCD00040990
7475563
XLBGYZLICFMDDS-UHFFFAOYSA-N
propan-2-yl 4,4,4-trifluoro-3-oxobutanoate
586897
95%

Chemical Identifiers

175230-50-9
198.14
XLBGYZLICFMDDS-UHFFFAOYSA-N
586897
CC(C)OC(=O)CC(=O)C(F)(F)F
C7H9F3O3
MFCD00040990
isopropyl 4,4,4-trifluoroacetoacetate, isopropyl 4,4,4-trifluoro-3-oxobutanoate, isopropyl 4,4,4-trifluoro-3-oxobutyrate, isopropyl-4,4,4-trifluoroacetoacetate, acmc-209e9i, isopropyl4,4,4-trifluoroacetoacetate, methylethyl 4,4,4-trifluoro-3-oxobutanoate, isopropyl 4,4,4-trifluoro-3-oxobutanoate #, 4,4,4-trifluoroacetoacetic acid isopropyl ester
propan-2-yl 4,4,4-trifluoro-3-oxobutanoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H271-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions